Assay Design



Gene Transcripts

Gene Symbol
ZNRF3
Entrez Gene ID
84133
Gene Name
zinc and ring finger 3
Gene Aliases
BK747E2.3, RNF203
Chromosome Location
Chr.22: 28800657 - 29057488 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206998.1 NP_001193927.1 4 - 5 632 71
NM_032173.3 NP_115549.2 4 - 5 425 71
XM_011530435.2 XP_011528737.1 5 - 6 621 71
XM_011530436.2 XP_011528738.1 6 - 7 676 71
XM_011530437.1 XP_011528739.1 5 - 6 1346 71
XM_011530438.2 XP_011528740.1 5 - 6 862 71
XM_017028990.1 XP_016884479.1 5 - 6 1117 71
GenBank mRNA AB051436.1 - 4 - 5 497 71
AK096397.1 - 4 - 5 425 71
CR456397.1 - 4 - 5 405 71

View Full Product Details