Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38414673-38415164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR1
Entrez Gene ID
2260
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
UniGene
Hs.264887
Gene Chromosome Location
Chr.8: 38411138-38468834 on Build GRCh38
Transcripts
NM_001174063.1 NM_001174064.1 NM_001174065.1 NM_001174066.1 NM_001174067.1 NM_015850.3 NM_023105.2 NM_023106.2 NM_023110.2 XM_006716303.2 XM_006716304.1 XM_006716306.2 XM_006716307.1 XM_006716309.3 XM_006716310.2 XM_006716311.1 XM_006716312.1 XM_006716313.2 XM_006716314.1 XM_011544443.2 XM_011544444.1 XM_011544445.2 XM_011544446.2 XM_011544447.2 XM_011544448.1 XM_011544449.1 XM_011544450.2 XM_011544451.1 XM_017013219.1 XM_017013220.1 XM_017013221.1 XM_017013222.1 XM_017013223.1 XM_017013224.1 XM_017013225.1 XM_017013226.1 XM_017013227.1 XM_017013228.1 XM_017013229.1 XM_017013230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6578117 c.1968G>C p.K656N Chr.8: 38414788-38414788 on Build GRCh38
1137867 c.1855G>C p.V619L Chr.8: 38414871-38414871 on Build GRCh38
486436 c.1885G>C p.V629L Chr.8: 38414871-38414871 on Build GRCh38
4445156 c.1954G>A p.D652N Chr.8: 38414802-38414802 on Build GRCh38
5874658 c.1906C>T p.R636W Chr.8: 38414820-38414820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901904245 Chr.8: 38414963-38414963 on Build GRCh38
rs4647905 Chr.8: 38415024-38415024 on Build GRCh38
rs765717436 Chr.8: 38414942-38414942 on Build GRCh38
rs1029181940 Chr.8: 38414728-38414728 on Build GRCh38
rs370630198 Chr.8: 38414938-38414938 on Build GRCh38
rs145273148 Chr.8: 38414739-38414739 on Build GRCh38
rs962335970 Chr.8: 38414986-38414986 on Build GRCh38
rs1006427079 Chr.8: 38414984-38414984 on Build GRCh38
rs561125538 Chr.8: 38414997-38414997 on Build GRCh38
rs746473852 Chr.8: 38414723-38414723 on Build GRCh38
rs760875320 Chr.8: 38415047-38415047 on Build GRCh38
rs942166487 Chr.8: 38415127-38415127 on Build GRCh38
rs727505370 Chr.8: 38414840-38414840 on Build GRCh38
rs4647903 Chr.8: 38415064-38415064 on Build GRCh38
rs770743973 Chr.8: 38414923-38414923 on Build GRCh38
rs773947414 Chr.8: 38414924-38414924 on Build GRCh38
rs983615443 Chr.8: 38414929-38414929 on Build GRCh38
rs778015347 Chr.8: 38414732-38414732 on Build GRCh38
rs869025672 Chr.8: 38414872-38414872 on Build GRCh38
rs764253969 Chr.8: 38414800-38414800 on Build GRCh38
rs1057518481 Chr.8: 38414903-38414903 on Build GRCh38
rs766962343 Chr.8: 38414812-38414812 on Build GRCh38
rs1039466396 Chr.8: 38415123-38415123 on Build GRCh38
rs753193082 Chr.8: 38414820-38414820 on Build GRCh38
rs776590352 Chr.8: 38414760-38414760 on Build GRCh38
rs746123129 Chr.8: 38414868-38414868 on Build GRCh38
rs769572642 Chr.8: 38414764-38414764 on Build GRCh38
rs917323541 Chr.8: 38414701-38414701 on Build GRCh38
rs876661334 Chr.8: 38414778-38414778 on Build GRCh38
rs1050495737 Chr.8: 38415088-38415088 on Build GRCh38
rs996242692 Chr.8: 38414966-38414966 on Build GRCh38
rs756586457 Chr.8: 38414824-38414824 on Build GRCh38
rs987999400 Chr.8: 38415040-38415040 on Build GRCh38
rs757656065 Chr.8: 38414940-38414940 on Build GRCh38
rs577599441 Chr.8: 38414798-38414798 on Build GRCh38
rs869217053 Chr.8: 38414724-38414725 on Build GRCh38
rs368768306 Chr.8: 38415066-38415066 on Build GRCh38
rs745989679 Chr.8: 38414752-38414752 on Build GRCh38
rs368389787 Chr.8: 38414931-38414931 on Build GRCh38
rs121909628 Chr.8: 38414892-38414892 on Build GRCh38
rs768446923 Chr.8: 38414757-38414757 on Build GRCh38
rs771342533 Chr.8: 38414746-38414746 on Build GRCh38
rs932002120 Chr.8: 38415080-38415080 on Build GRCh38
rs869025671 Chr.8: 38414876-38414876 on Build GRCh38
rs398122946 Chr.8: 38414889-38414889 on Build GRCh38
rs761228427 Chr.8: 38414932-38414932 on Build GRCh38
rs767581987 Chr.8: 38414993-38414993 on Build GRCh38
rs1050527237 Chr.8: 38414967-38414967 on Build GRCh38
rs869320694 Chr.8: 38414790-38414790 on Build GRCh38
rs542564959 Chr.8: 38414972-38414972 on Build GRCh38
rs779120005 Chr.8: 38414749-38414749 on Build GRCh38
rs376421301 Chr.8: 38414803-38414803 on Build GRCh38
rs780009859 Chr.8: 38414887-38414887 on Build GRCh38
rs769679933 Chr.8: 38414914-38414914 on Build GRCh38
rs983971417 Chr.8: 38414699-38414699 on Build GRCh38
rs762104979 Chr.8: 38414985-38414985 on Build GRCh38
rs544967630 Chr.8: 38414852-38414852 on Build GRCh38
rs927214668 Chr.8: 38414736-38414736 on Build GRCh38
rs876661333 Chr.8: 38414828-38414828 on Build GRCh38
rs923252249 Chr.8: 38415070-38415070 on Build GRCh38
rs564942100 Chr.8: 38415043-38415043 on Build GRCh38
rs1036552478 Chr.8: 38415135-38415135 on Build GRCh38
rs748049385 Chr.8: 38414909-38414909 on Build GRCh38
rs958071759 Chr.8: 38415027-38415027 on Build GRCh38
rs543518207 Chr.8: 38415026-38415026 on Build GRCh38
rs148750337 Chr.8: 38414763-38414763 on Build GRCh38
rs371160786 Chr.8: 38414917-38414917 on Build GRCh38
rs751038400 Chr.8: 38414802-38414802 on Build GRCh38
rs751366805 Chr.8: 38414912-38414915 on Build GRCh38
rs946004304 Chr.8: 38414957-38414957 on Build GRCh38
rs778191968 Chr.8: 38414836-38414836 on Build GRCh38
rs969056825 Chr.8: 38415013-38415013 on Build GRCh38
rs201805975 Chr.8: 38414761-38414761 on Build GRCh38
rs146284363 Chr.8: 38415136-38415136 on Build GRCh38
rs749452174 Chr.8: 38414741-38414741 on Build GRCh38
rs754309928 Chr.8: 38414851-38414851 on Build GRCh38
rs1024580232 Chr.8: 38415002-38415002 on Build GRCh38
rs375014194 Chr.8: 38414925-38414925 on Build GRCh38
rs762777083 Chr.8: 38414919-38414919 on Build GRCh38
rs772140963 Chr.8: 38414881-38414881 on Build GRCh38
rs887877444 Chr.8: 38415089-38415089 on Build GRCh38
rs770318743 Chr.8: 38414769-38414769 on Build GRCh38
rs372632166 Chr.8: 38414770-38414770 on Build GRCh38
rs1040410463 Chr.8: 38414962-38414962 on Build GRCh38

View Full Product Details