Assay Design



Gene Transcripts

Gene Symbol
RAB3A
Entrez Gene ID
5864
Gene Name
RAB3A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.19: 18196799 - 18204064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002866.4 NP_002857.1 1 - 2 172 62
GenBank mRNA AF254795.1 - 1 - 2 132 62
AK289559.1 - 1 - 2 150 62
AK293467.1 - - 146 62
BC011782.2 - 1 - 2 137 62
DB497194.1 - 1 - 2 182 62

View Full Product Details