Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258312.1 | NP_001245241.1 | 14 - 15 | 1779 | 116 |
NM_005019.4 | NP_005010.2 | 13 - 14 | 1794 | 116 | |
XM_017004295.1 | XP_016859784.1 | 14 - 15 | 1799 | 116 | |
XM_017004296.1 | XP_016859785.1 | 14 - 15 | 1721 | 116 | |
XM_017004297.1 | XP_016859786.1 | 14 - 15 | 1662 | 116 | |
XM_017004298.1 | XP_016859787.1 | 14 - 15 | 1844 | 116 | |
XM_017004302.1 | XP_016859791.1 | 13 - 14 | 1926 | 116 | |
GenBank mRNA | AF110238.1 | - | 2 - 3 | 207 | 116 |
AK294239.1 | - | 14 - 15 | 1714 | 116 | |
AK295657.1 | - | 14 - 15 | 1584 | 116 | |
BC022480.1 | - | 13 - 14 | 1762 | 116 |