Assay Design
Gene Transcripts
- Gene Symbol
- RGS14
- Entrez Gene ID
-
10636
- Gene Name
- regulator of G-protein signaling 14
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 177357671 - 177372601 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_006480.4 |
NP_006471.2 |
9 - 10 |
1236 |
75 |
GenBank mRNA |
BC014094.2 |
- |
9 - 10 |
1206 |
75 |
View Full Product Details