Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006499.4 | NP_006490.3 | 3 - 4 | 426 | 117 |
NM_201543.2 | NP_963837.1 | 3 - 4 | 426 | 117 | |
NM_201544.2 | NP_963838.1 | 2 - 3 | 483 | 117 | |
NM_201545.2 | NP_963839.1 | 3 - 4 | 296 | 117 | |
XM_011544188.2 | XP_011542490.1 | 2 - 3 | 240 | 117 | |
XM_017001274.1 | XP_016856763.1 | 2 - 3 | 240 | 117 | |
XM_017001275.1 | XP_016856764.1 | 2 - 3 | 240 | 117 | |
GenBank mRNA | AB209914.1 | - | 1 - 2 | 117 | |
AF074000.1 | - | 3 - 4 | 117 | ||
AF074001.1 | - | 3 - 4 | 117 | ||
AF074002.1 | - | 3 - 4 | 117 | ||
AF342815.1 | - | 1 - 2 | 71 | 117 | |
AF342816.1 | - | 1 - 2 | 71 | 117 | |
AK098613.1 | - | 2 - 3 | 117 | ||
AK296061.1 | - | 2 - 3 | 192 | 117 | |
AU117713.1 | - | 3 - 4 | 117 | ||
BC015818.1 | - | 2 - 3 | 117 | ||
BC016486.2 | - | 2 - 3 | 433 | 117 | |
BP280070.1 | - | 2 - 3 | 117 | ||
BP371701.1 | - | 4 - 5 | 117 | ||
BU561073.1 | - | 3 - 4 | 247 | 117 | |
L78132.1 | - | 1 - 2 | 98 | 117 | |
X91790.1 | - | 1 - 2 | 84 | 117 |