Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 1782063-1782598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN8
Entrez Gene ID
2055
Gene Name
ceroid-lipofuscinosis, neuronal 8
Gene Aliases
C8orf61, EPMR
UniGene
Hs.127675
Gene Chromosome Location
Chr.8: 1755778-1786570 on Build GRCh38
Transcripts
NM_018941.3

SNP Detail

SNP ID Chromosome Location
rs548356940 Chr.8: 1782497-1782497 on Build GRCh38
rs542709018 Chr.8: 1782371-1782371 on Build GRCh38
rs987354020 Chr.8: 1782125-1782125 on Build GRCh38
rs538670235 Chr.8: 1782240-1782240 on Build GRCh38
rs895953295 Chr.8: 1782195-1782195 on Build GRCh38
rs918194675 Chr.8: 1782286-1782286 on Build GRCh38
rs958054050 Chr.8: 1782091-1782092 on Build GRCh38
rs1041894827 Chr.8: 1782503-1782503 on Build GRCh38
rs938217427 Chr.8: 1782265-1782265 on Build GRCh38
rs1008723915 Chr.8: 1782550-1782550 on Build GRCh38
rs115698196 Chr.8: 1782529-1782529 on Build GRCh38
rs945650861 Chr.8: 1782292-1782292 on Build GRCh38
rs1056684132 Chr.8: 1782109-1782109 on Build GRCh38
rs370222132 Chr.8: 1782488-1782488 on Build GRCh38
rs902909884 Chr.8: 1782154-1782154 on Build GRCh38
rs888948903 Chr.8: 1782168-1782168 on Build GRCh38
rs920678882 Chr.8: 1782155-1782156 on Build GRCh38
rs567865373 Chr.8: 1782505-1782505 on Build GRCh38
rs536360279 Chr.8: 1782184-1782184 on Build GRCh38
rs985286383 Chr.8: 1782214-1782214 on Build GRCh38
rs78155116 Chr.8: 1782528-1782528 on Build GRCh38
rs1040200609 Chr.8: 1782562-1782562 on Build GRCh38
rs567332574 Chr.8: 1782155-1782155 on Build GRCh38
rs924297821 Chr.8: 1782498-1782498 on Build GRCh38
rs769540779 Chr.8: 1782450-1782450 on Build GRCh38
rs935698172 Chr.8: 1782506-1782506 on Build GRCh38
rs571941797 Chr.8: 1782090-1782090 on Build GRCh38
rs111709829 Chr.8: 1782296-1782296 on Build GRCh38
rs987324210 Chr.8: 1782403-1782403 on Build GRCh38
rs142012947 Chr.8: 1782366-1782366 on Build GRCh38
rs992359255 Chr.8: 1782273-1782273 on Build GRCh38
rs991494035 Chr.8: 1782491-1782491 on Build GRCh38
rs1014815335 Chr.8: 1782424-1782424 on Build GRCh38
rs1041927739 Chr.8: 1782202-1782202 on Build GRCh38
rs143679376 Chr.8: 1782116-1782116 on Build GRCh38
rs180870105 Chr.8: 1782326-1782326 on Build GRCh38
rs938228748 Chr.8: 1782102-1782102 on Build GRCh38
rs563039664 Chr.8: 1782420-1782420 on Build GRCh38
rs748731042 Chr.8: 1782575-1782575 on Build GRCh38
rs758411737 Chr.8: 1782097-1782097 on Build GRCh38
rs374346831 Chr.8: 1782200-1782200 on Build GRCh38
rs937447613 Chr.8: 1782169-1782169 on Build GRCh38
rs999185479 Chr.8: 1782239-1782239 on Build GRCh38
rs73541034 Chr.8: 1782527-1782527 on Build GRCh38
rs148153870 Chr.8: 1782237-1782237 on Build GRCh38
rs950189875 Chr.8: 1782499-1782499 on Build GRCh38
rs952133556 Chr.8: 1782213-1782213 on Build GRCh38
rs528003128 Chr.8: 1782446-1782446 on Build GRCh38
rs565558303 Chr.8: 1782148-1782148 on Build GRCh38
rs972080269 Chr.8: 1782190-1782190 on Build GRCh38
rs903444299 Chr.8: 1782238-1782238 on Build GRCh38
rs779844295 Chr.8: 1782167-1782167 on Build GRCh38
rs901683560 Chr.8: 1782571-1782571 on Build GRCh38
rs531591559 Chr.8: 1782421-1782421 on Build GRCh38
rs949752324 Chr.8: 1782132-1782132 on Build GRCh38
rs920632202 Chr.8: 1782445-1782445 on Build GRCh38
rs377434706 Chr.8: 1782441-1782441 on Build GRCh38
rs952184350 Chr.8: 1782437-1782437 on Build GRCh38
rs775467130 Chr.8: 1782559-1782559 on Build GRCh38
rs1054651324 Chr.8: 1782312-1782312 on Build GRCh38
rs796537466 Chr.8: 1782201-1782201 on Build GRCh38
rs771972051 Chr.8: 1782539-1782539 on Build GRCh38
rs936288901 Chr.8: 1782521-1782521 on Build GRCh38
rs918098434 Chr.8: 1782451-1782451 on Build GRCh38
rs888944303 Chr.8: 1782322-1782322 on Build GRCh38
rs890222920 Chr.8: 1782543-1782543 on Build GRCh38
rs536757746 Chr.8: 1782440-1782440 on Build GRCh38
rs62476924 Chr.8: 1782185-1782185 on Build GRCh38
rs935725111 Chr.8: 1782306-1782306 on Build GRCh38
rs560764646 Chr.8: 1782361-1782361 on Build GRCh38
rs755799935 Chr.8: 1782347-1782347 on Build GRCh38
rs1049203000 Chr.8: 1782094-1782094 on Build GRCh38
rs572334654 Chr.8: 1782302-1782302 on Build GRCh38
rs750735822 Chr.8: 1782170-1782170 on Build GRCh38
rs945600082 Chr.8: 1782482-1782482 on Build GRCh38
rs780452402 Chr.8: 1782225-1782225 on Build GRCh38
rs532802873 Chr.8: 1782175-1782175 on Build GRCh38
rs754831539 Chr.8: 1782285-1782285 on Build GRCh38
rs1053450000 Chr.8: 1782522-1782522 on Build GRCh38

View Full Product Details