Assay Design



Gene Transcripts

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
Chromosome Location
Chr.17: 60677811 - 61392838 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099432.1 NP_001092902.1 11 - 12 930 83
NM_001320470.1 NP_001307399.1 11 - 12 930 83
NM_017679.3 NP_060149.3 11 - 12 930 83
XM_005257472.1 XP_005257529.1 11 - 12 881 83
XM_011524939.1 XP_011523241.1 11 - 12 881 83
XM_011524940.1 XP_011523242.1 11 - 12 881 83
XM_011524941.1 XP_011523243.1 11 - 12 881 83
XM_011524942.2 XP_011523244.1 11 - 12 881 83
XM_011524943.2 XP_011523245.1 11 - 12 881 83
XM_011524944.1 XP_011523246.1 11 - 12 881 83
XM_011524945.2 XP_011523247.1 11 - 12 881 83
XM_011524946.2 XP_011523248.1 11 - 12 881 83
XM_011524947.1 XP_011523249.1 11 - 12 881 83
XM_011524948.2 XP_011523250.1 11 - 12 881 83
XM_011524949.2 XP_011523251.1 11 - 12 881 83
XM_011524950.1 XP_011523252.1 11 - 12 881 83
XM_011524951.2 XP_011523253.1 11 - 12 881 83
XM_017024783.1 XP_016880272.1 11 - 12 881 83
XM_017024784.1 XP_016880273.1 11 - 12 881 83
XM_017024785.1 XP_016880274.1 11 - 12 881 83
XM_017024786.1 XP_016880275.1 11 - 12 881 83
XM_017024787.1 XP_016880276.1 11 - 12 881 83
XM_017024788.1 XP_016880277.1 11 - 12 881 83
XM_017024789.1 XP_016880278.1 11 - 12 881 83
XM_017024790.1 XP_016880279.1 11 - 12 881 83
XM_017024791.1 XP_016880280.1 11 - 12 881 83
XM_017024792.1 XP_016880281.1 11 - 12 881 83
XM_017024793.1 XP_016880282.1 11 - 12 881 83
XM_017024794.1 XP_016880283.1 11 - 12 881 83
XM_017024795.1 XP_016880284.1 11 - 12 881 83
XM_017024796.1 XP_016880285.1 3 - 4 307 83
GenBank mRNA AF361219.1 - 12 - 13 869 83
AJ518105.1 - 3 - 4 322 83
AJ518836.1 - 3 - 4 237 83
AK022526.1 - 3 - 4 285 83
AK023054.1 - 3 - 4 277 83
AK025510.1 - 11 - 12 846 83
AK225757.1 - 11 - 12 889 83
AK314744.1 - 11 - 12 875 83
BC017390.1 - 11 - 12 886 83
BC117275.1 - 11 - 12 852 83
BC143386.1 - 11 - 12 852 83
BC143387.1 - 11 - 12 852 83

View Full Product Details