Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001740.3 | NP_001001740.1 | 8 - 9 | 1218 | 123 |
NM_001286644.1 | NP_001273573.1 | 7 - 8 | 1093 | 123 | |
NM_022457.6 | NP_071902.2 | 9 - 10 | 1290 | 123 | |
XM_005245447.2 | XP_005245504.1 | 9 - 10 | 1313 | 123 | |
XM_005245448.2 | XP_005245505.1 | 8 - 9 | 1265 | 123 | |
XM_006711487.2 | XP_006711550.1 | 8 - 9 | 1256 | 123 | |
XM_017002061.1 | XP_016857550.1 | 9 - 10 | 1325 | 123 | |
XM_017002063.1 | XP_016857552.1 | 9 - 10 | 1313 | 123 | |
XM_017002065.1 | XP_016857554.1 | 8 - 9 | 1265 | 123 | |
XM_017002066.1 | XP_016857555.1 | 8 - 9 | 1256 | 123 | |
XM_017002067.1 | XP_016857556.1 | 8 - 9 | 1253 | 123 | |
XM_017002075.1 | XP_016857564.1 | 6 - 7 | 494 | 123 | |
XM_017002076.1 | XP_016857565.1 | 7 - 8 | 477 | 123 | |
GenBank mRNA | AF508940.1 | - | 9 - 10 | 1024 | 123 |
AF527539.1 | - | 9 - 10 | 1287 | 123 | |
AK001278.1 | - | 7 - 8 | 529 | 123 | |
AK314750.1 | - | 9 - 10 | 1222 | 123 | |
AY509921.1 | - | 8 - 9 | 952 | 123 | |
BC039723.1 | - | 7 - 8 | 540 | 123 | |
BC094728.1 | - | 9 - 10 | 1278 | 123 | |
KJ534928.1 | - | 9 - 10 | 1024 | 123 | |
KJ534930.1 | - | 8 - 9 | 964 | 123 | |
KJ534931.1 | - | 8 - 9 | 955 | 123 | |
KJ534932.1 | - | 9 - 10 | 1012 | 123 | |
KJ535061.1 | - | 6 - 7 | 729 | 123 | |
KJ535076.1 | - | 8 - 9 | 926 | 123 |