Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128843.2 | NP_001122315.2 | 6 - 7 | 484 | 82 |
NM_001244764.1 | NP_001231693.1 | 6 - 7 | 524 | 82 | |
NM_024873.5 | NP_079149.3 | 3 - 4 | 429 | 82 | |
XM_011532256.2 | XP_011530558.1 | 7 - 8 | 757 | 82 | |
XM_011532257.2 | XP_011530559.1 | 7 - 8 | 757 | 82 | |
XM_017008619.1 | XP_016864108.1 | 7 - 8 | 757 | 82 | |
XM_017008620.1 | XP_016864109.1 | 7 - 8 | 620 | 82 | |
XM_017008621.1 | XP_016864110.1 | 6 - 7 | 667 | 82 | |
XM_017008622.1 | XP_016864111.1 | 6 - 7 | 920 | 82 | |
XM_017008623.1 | XP_016864112.1 | 6 - 7 | 530 | 82 | |
XM_017008624.1 | XP_016864113.1 | 5 - 6 | 475 | 82 | |
XM_017008625.1 | XP_016864114.1 | 4 - 5 | 1008 | 82 | |
GenBank mRNA | AF277289.1 | - | 3 - 4 | 418 | 82 |
AJ320534.1 | - | 3 - 4 | 418 | 82 | |
AK290409.1 | - | 3 - 4 | 410 | 82 | |
AK301058.1 | - | 6 - 7 | 484 | 82 | |
AK303192.1 | - | 6 - 7 | 526 | 82 | |
BC128408.1 | - | 3 - 4 | 417 | 82 | |
BP335455.1 | - | 3 - 4 | 429 | 82 | |
DC389224.1 | - | 6 - 7 | 484 | 82 | |
EF585496.1 | - | 3 - 4 | 418 | 82 |