Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294340.1 | NP_001281269.1 | 1 - 2 | 186 | 105 |
NM_144604.3 | NP_653205.3 | 1 - 2 | 186 | 105 | |
XM_011522863.1 | XP_011521165.1 | 1 - 2 | 186 | 105 | |
XM_017022934.1 | XP_016878423.1 | 1 - 2 | 186 | 105 | |
GenBank mRNA | AK225807.1 | - | 1 - 2 | 156 | 105 |
AK300254.1 | - | 1 - 2 | 164 | 105 | |
AK302716.1 | - | 1 - 2 | 164 | 105 | |
BC050463.1 | - | 1 - 2 | 133 | 105 | |
CN299707.1 | - | 1 - 2 | 145 | 105 | |
DA313229.1 | - | 1 - 2 | 186 | 105 |