Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144658.3 | NP_653259.3 | 32 - 33 | 3654 | 88 |
XM_005262368.3 | XP_005262425.1 | 33 - 34 | 3938 | 88 | |
XM_005262369.4 | XP_005262426.1 | 33 - 34 | 3632 | 88 | |
XM_011531276.2 | XP_011529578.1 | 33 - 34 | 3573 | 88 | |
XM_011531278.2 | XP_011529580.1 | 20 - 21 | 2059 | 88 | |
GenBank mRNA | AK056684.1 | - | 2 - 3 | 168 | 88 |
AK124735.1 | - | 27 - 28 | 3077 | 88 | |
AK125641.1 | - | 11 - 12 | 1166 | 88 | |
AY692226.1 | - | 32 - 33 | 3654 | 88 |