Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 63059666-63060216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTPA
Entrez Gene ID
7274
Gene Name
alpha tocopherol transfer protein
Gene Aliases
ATTP, AVED, TTP1, alphaTTP
UniGene
Hs.69049
Gene Chromosome Location
Chr.8: 63058409-63086523 on Build GRCh38
Transcripts
NM_000370.3 XM_006716468.3

SNP Detail

SNP ID Chromosome Location
rs775811621 Chr.8: 63059865-63059865 on Build GRCh38
rs762967547 Chr.8: 63060000-63060000 on Build GRCh38
rs531791721 Chr.8: 63060136-63060136 on Build GRCh38
rs924033689 Chr.8: 63059803-63059803 on Build GRCh38
rs1038838681 Chr.8: 63059890-63059890 on Build GRCh38
rs886063060 Chr.8: 63059858-63059858 on Build GRCh38
rs139507839 Chr.8: 63060135-63060135 on Build GRCh38
rs547533075 Chr.8: 63059707-63059707 on Build GRCh38
rs535310725 Chr.8: 63059766-63059766 on Build GRCh38
rs979812877 Chr.8: 63060030-63060030 on Build GRCh38
rs950995318 Chr.8: 63059710-63059710 on Build GRCh38
rs968487520 Chr.8: 63059956-63059956 on Build GRCh38
rs567615411 Chr.8: 63060041-63060041 on Build GRCh38
rs6472068 Chr.8: 63059765-63059765 on Build GRCh38
rs942961912 Chr.8: 63060117-63060117 on Build GRCh38
rs934240987 Chr.8: 63059836-63059836 on Build GRCh38
rs1014173942 Chr.8: 63059964-63059964 on Build GRCh38
rs4587328 Chr.8: 63060159-63060159 on Build GRCh38
rs1024238503 Chr.8: 63059974-63059974 on Build GRCh38
rs553853197 Chr.8: 63059884-63059884 on Build GRCh38
rs188157484 Chr.8: 63059742-63059742 on Build GRCh38
rs550802288 Chr.8: 63059948-63059948 on Build GRCh38
rs779524525 Chr.8: 63060023-63060023 on Build GRCh38
rs913392511 Chr.8: 63060145-63060145 on Build GRCh38
rs969785751 Chr.8: 63059784-63059784 on Build GRCh38
rs886063062 Chr.8: 63060117-63060117 on Build GRCh38
rs1051064231 Chr.8: 63059814-63059814 on Build GRCh38
rs773557348 Chr.8: 63059768-63059771 on Build GRCh38
rs913423928 Chr.8: 63059830-63059830 on Build GRCh38
rs980227738 Chr.8: 63059788-63059788 on Build GRCh38
rs898592464 Chr.8: 63059914-63059914 on Build GRCh38
rs958888424 Chr.8: 63059760-63059760 on Build GRCh38
rs889749725 Chr.8: 63059876-63059876 on Build GRCh38
rs577649057 Chr.8: 63059828-63059828 on Build GRCh38
rs912576648 Chr.8: 63060116-63060116 on Build GRCh38
rs986805689 Chr.8: 63060109-63060109 on Build GRCh38
rs1026134648 Chr.8: 63059938-63059938 on Build GRCh38
rs150991402 Chr.8: 63059737-63059737 on Build GRCh38
rs951129782 Chr.8: 63059907-63059907 on Build GRCh38
rs995683122 Chr.8: 63059851-63059851 on Build GRCh38
rs916917522 Chr.8: 63059778-63059778 on Build GRCh38
rs181348477 Chr.8: 63059887-63059887 on Build GRCh38
rs866321408 Chr.8: 63059921-63059921 on Build GRCh38
rs75095572 Chr.8: 63059768-63059768 on Build GRCh38
rs56675911 Chr.8: 63059848-63059848 on Build GRCh38
rs758747715 Chr.8: 63060185-63060185 on Build GRCh38
rs958631985 Chr.8: 63059947-63059947 on Build GRCh38
rs934167634 Chr.8: 63060146-63060146 on Build GRCh38
rs565546241 Chr.8: 63060164-63060164 on Build GRCh38
rs924002477 Chr.8: 63060002-63060002 on Build GRCh38
rs957122787 Chr.8: 63060111-63060111 on Build GRCh38
rs553317836 Chr.8: 63059811-63059811 on Build GRCh38
rs970090396 Chr.8: 63060011-63060011 on Build GRCh38
rs942666781 Chr.8: 63060180-63060180 on Build GRCh38
rs542746881 Chr.8: 63060021-63060021 on Build GRCh38
rs925637194 Chr.8: 63060042-63060042 on Build GRCh38
rs886063061 Chr.8: 63060088-63060088 on Build GRCh38
rs548084272 Chr.8: 63060071-63060073 on Build GRCh38
rs778021506 Chr.8: 63060188-63060188 on Build GRCh38
rs537396248 Chr.8: 63059789-63059789 on Build GRCh38

View Full Product Details