Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 63065814-63066319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTPA
Entrez Gene ID
7274
Gene Name
alpha tocopherol transfer protein
Gene Aliases
ATTP, AVED, TTP1, alphaTTP
UniGene
Hs.69049
Gene Chromosome Location
Chr.8: 63058409-63086523 on Build GRCh38
Transcripts
NM_000370.3 XM_006716468.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1100995 c.393C>T p.D131D Chr.8: 63066063-63066063 on Build GRCh38
5409759 c.487T>C p.W163R Chr.8: 63065969-63065969 on Build GRCh38
7311895 c.460A>G p.I154V Chr.8: 63065996-63065996 on Build GRCh38
1100993 c.424C>A p.L142I Chr.8: 63066032-63066032 on Build GRCh38
3900940 c.409C>T p.L137L Chr.8: 63066047-63066047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113600979 Chr.8: 63066131-63066131 on Build GRCh38
rs553783588 Chr.8: 63065917-63065917 on Build GRCh38
rs755714678 Chr.8: 63065991-63065991 on Build GRCh38
rs780098228 Chr.8: 63066121-63066121 on Build GRCh38
rs375011573 Chr.8: 63065998-63065998 on Build GRCh38
rs555775890 Chr.8: 63066062-63066062 on Build GRCh38
rs777822041 Chr.8: 63066143-63066143 on Build GRCh38
rs756867504 Chr.8: 63066021-63066021 on Build GRCh38
rs397515525 Chr.8: 63065908-63065908 on Build GRCh38
rs373811993 Chr.8: 63066054-63066054 on Build GRCh38
rs895383170 Chr.8: 63066161-63066161 on Build GRCh38
rs150710403 Chr.8: 63066004-63066004 on Build GRCh38
rs377115359 Chr.8: 63065945-63065945 on Build GRCh38
rs886040964 Chr.8: 63065902-63065902 on Build GRCh38
rs973916151 Chr.8: 63066005-63066005 on Build GRCh38
rs775410361 Chr.8: 63065953-63065953 on Build GRCh38
rs34090391 Chr.8: 63066196-63066196 on Build GRCh38
rs777536431 Chr.8: 63065993-63065993 on Build GRCh38
rs34258920 Chr.8: 63066063-63066063 on Build GRCh38
rs866773722 Chr.8: 63065968-63065968 on Build GRCh38
rs202136581 Chr.8: 63065905-63065905 on Build GRCh38
rs544337864 Chr.8: 63066198-63066198 on Build GRCh38
rs1013615578 Chr.8: 63066274-63066274 on Build GRCh38
rs201788259 Chr.8: 63066100-63066100 on Build GRCh38
rs763622589 Chr.8: 63066098-63066098 on Build GRCh38
rs768748858 Chr.8: 63065992-63065992 on Build GRCh38
rs767879332 Chr.8: 63065978-63065978 on Build GRCh38
rs779719601 Chr.8: 63066042-63066042 on Build GRCh38
rs933007891 Chr.8: 63065861-63065861 on Build GRCh38
rs778952721 Chr.8: 63066023-63066023 on Build GRCh38
rs367736684 Chr.8: 63065866-63065866 on Build GRCh38
rs138483857 Chr.8: 63066080-63066080 on Build GRCh38
rs752986314 Chr.8: 63066079-63066079 on Build GRCh38
rs929504887 Chr.8: 63066258-63066258 on Build GRCh38
rs181109321 Chr.8: 63065904-63065904 on Build GRCh38
rs753557931 Chr.8: 63066103-63066103 on Build GRCh38
rs201891788 Chr.8: 63066108-63066108 on Build GRCh38
rs773161117 Chr.8: 63065946-63065946 on Build GRCh38
rs761956631 Chr.8: 63065941-63065941 on Build GRCh38
rs745820087 Chr.8: 63066028-63066028 on Build GRCh38
rs200236627 Chr.8: 63065886-63065886 on Build GRCh38
rs34647756 Chr.8: 63065942-63065942 on Build GRCh38
rs748164236 Chr.8: 63065969-63065969 on Build GRCh38
rs754846253 Chr.8: 63066134-63066134 on Build GRCh38
rs746890193 Chr.8: 63066045-63066045 on Build GRCh38
rs1057517448 Chr.8: 63066015-63066015 on Build GRCh38
rs745683556 Chr.8: 63065882-63065885 on Build GRCh38
rs752104187 Chr.8: 63065972-63065972 on Build GRCh38
rs1022945090 Chr.8: 63065987-63065987 on Build GRCh38
rs760038401 Chr.8: 63066070-63066070 on Build GRCh38
rs759856643 Chr.8: 63065975-63065975 on Build GRCh38
rs397515378 Chr.8: 63065970-63065970 on Build GRCh38
rs397515524 Chr.8: 63066035-63066035 on Build GRCh38
rs533373396 Chr.8: 63066265-63066265 on Build GRCh38
rs121917851 Chr.8: 63066056-63066056 on Build GRCh38
rs758406902 Chr.8: 63066117-63066117 on Build GRCh38
rs772020906 Chr.8: 63065934-63065934 on Build GRCh38
rs572263038 Chr.8: 63065985-63065985 on Build GRCh38
rs776942277 Chr.8: 63065939-63065939 on Build GRCh38
rs398123340 Chr.8: 63066100-63066100 on Build GRCh38
rs764851051 Chr.8: 63066113-63066113 on Build GRCh38
rs779038537 Chr.8: 63065930-63065930 on Build GRCh38
rs758413130 Chr.8: 63066246-63066246 on Build GRCh38
rs562610976 Chr.8: 63066248-63066248 on Build GRCh38
rs17854960 Chr.8: 63066078-63066078 on Build GRCh38
rs202246894 Chr.8: 63066108-63066108 on Build GRCh38
rs745623765 Chr.8: 63065933-63065933 on Build GRCh38
rs397515379 Chr.8: 63065942-63065943 on Build GRCh38
rs537986447 Chr.8: 63065873-63065873 on Build GRCh38
rs766942596 Chr.8: 63065864-63065864 on Build GRCh38
rs768075601 Chr.8: 63066075-63066075 on Build GRCh38
rs771937122 Chr.8: 63066036-63066036 on Build GRCh38
rs758837874 Chr.8: 63065863-63065863 on Build GRCh38
rs918456240 Chr.8: 63066018-63066018 on Build GRCh38
rs898038340 Chr.8: 63065943-63065943 on Build GRCh38
rs770760719 Chr.8: 63065927-63065927 on Build GRCh38
rs748038814 Chr.8: 63066055-63066055 on Build GRCh38
rs755385428 Chr.8: 63065870-63065870 on Build GRCh38
rs185200764 Chr.8: 63066255-63066255 on Build GRCh38
rs750321633 Chr.8: 63066114-63066114 on Build GRCh38
rs988427817 Chr.8: 63066053-63066053 on Build GRCh38
rs756777636 Chr.8: 63066107-63066107 on Build GRCh38
rs1057499096 Chr.8: 63066259-63066259 on Build GRCh38
rs947008685 Chr.8: 63066178-63066178 on Build GRCh38
rs766852761 Chr.8: 63065952-63065952 on Build GRCh38

View Full Product Details