Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 63072793-63073297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTPA
Entrez Gene ID
7274
Gene Name
alpha tocopherol transfer protein
Gene Aliases
ATTP, AVED, TTP1, alphaTTP
UniGene
Hs.69049
Gene Chromosome Location
Chr.8: 63058409-63086523 on Build GRCh38
Transcripts
NM_000370.3 XM_006716468.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3951767 c.205-5T>C p.? Chr.8: 63073093-63073093 on Build GRCh38
3374964 c.291G>T p.K97N Chr.8: 63073002-63073002 on Build GRCh38
338082 c.358+1G>T p.? Chr.8: 63072934-63072934 on Build GRCh38
6383438 c.245A>T p.E82V Chr.8: 63073048-63073048 on Build GRCh38
1100996 c.285C>T p.L95L Chr.8: 63073008-63073008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781777118 Chr.8: 63073115-63073115 on Build GRCh38
rs199925260 Chr.8: 63072903-63072903 on Build GRCh38
rs760779083 Chr.8: 63072906-63072907 on Build GRCh38
rs1000759000 Chr.8: 63073103-63073103 on Build GRCh38
rs776814829 Chr.8: 63073088-63073089 on Build GRCh38
rs553748670 Chr.8: 63073148-63073148 on Build GRCh38
rs758030471 Chr.8: 63072936-63072936 on Build GRCh38
rs200760380 Chr.8: 63072998-63072998 on Build GRCh38
rs371651447 Chr.8: 63073075-63073075 on Build GRCh38
rs1012996856 Chr.8: 63073073-63073073 on Build GRCh38
rs121917849 Chr.8: 63072990-63072990 on Build GRCh38
rs777447024 Chr.8: 63073107-63073107 on Build GRCh38
rs774406845 Chr.8: 63072890-63072890 on Build GRCh38
rs756753417 Chr.8: 63073213-63073213 on Build GRCh38
rs886040963 Chr.8: 63073089-63073089 on Build GRCh38
rs758349851 Chr.8: 63073090-63073090 on Build GRCh38
rs201341685 Chr.8: 63072904-63072904 on Build GRCh38
rs748000998 Chr.8: 63072964-63072964 on Build GRCh38
rs368131865 Chr.8: 63073072-63073072 on Build GRCh38
rs910610963 Chr.8: 63073253-63073253 on Build GRCh38
rs555448323 Chr.8: 63073185-63073185 on Build GRCh38
rs35505341 Chr.8: 63073176-63073176 on Build GRCh38
rs536080176 Chr.8: 63073031-63073031 on Build GRCh38
rs34162030 Chr.8: 63073157-63073157 on Build GRCh38
rs779151906 Chr.8: 63072979-63072979 on Build GRCh38
rs769549473 Chr.8: 63073015-63073015 on Build GRCh38
rs750665523 Chr.8: 63073082-63073082 on Build GRCh38
rs373191737 Chr.8: 63072901-63072901 on Build GRCh38
rs143010236 Chr.8: 63072935-63072935 on Build GRCh38
rs549121543 Chr.8: 63072827-63072827 on Build GRCh38
rs772224814 Chr.8: 63072984-63072984 on Build GRCh38
rs747563020 Chr.8: 63073001-63073001 on Build GRCh38
rs374483216 Chr.8: 63072942-63072942 on Build GRCh38
rs550046756 Chr.8: 63072983-63072983 on Build GRCh38
rs146136556 Chr.8: 63073014-63073014 on Build GRCh38
rs769194493 Chr.8: 63073110-63073110 on Build GRCh38
rs374272548 Chr.8: 63073028-63073028 on Build GRCh38
rs759510197 Chr.8: 63073034-63073034 on Build GRCh38
rs766200402 Chr.8: 63072938-63072938 on Build GRCh38
rs954442288 Chr.8: 63073270-63073270 on Build GRCh38
rs757454805 Chr.8: 63072965-63072965 on Build GRCh38
rs751549880 Chr.8: 63072941-63072941 on Build GRCh38
rs761564561 Chr.8: 63072921-63072921 on Build GRCh38
rs368391591 Chr.8: 63073002-63073002 on Build GRCh38
rs955354789 Chr.8: 63073183-63073183 on Build GRCh38
rs746001498 Chr.8: 63072896-63072896 on Build GRCh38
rs765051181 Chr.8: 63072928-63072928 on Build GRCh38
rs773405362 Chr.8: 63072907-63072907 on Build GRCh38
rs766675875 Chr.8: 63073073-63073074 on Build GRCh38
rs1057516423 Chr.8: 63072980-63072980 on Build GRCh38
rs985338429 Chr.8: 63073212-63073212 on Build GRCh38
rs764540742 Chr.8: 63072907-63072907 on Build GRCh38
rs371592549 Chr.8: 63072920-63072920 on Build GRCh38
rs770928018 Chr.8: 63072887-63072887 on Build GRCh38
rs200547556 Chr.8: 63072955-63072955 on Build GRCh38
rs376126799 Chr.8: 63072902-63072902 on Build GRCh38
rs767409948 Chr.8: 63073042-63073042 on Build GRCh38
rs987936542 Chr.8: 63072820-63072820 on Build GRCh38
rs910649572 Chr.8: 63072873-63072873 on Build GRCh38
rs186021365 Chr.8: 63073021-63073021 on Build GRCh38
rs749536567 Chr.8: 63072885-63072885 on Build GRCh38
rs768835088 Chr.8: 63073012-63073012 on Build GRCh38
rs780184310 Chr.8: 63073098-63073098 on Build GRCh38
rs752463700 Chr.8: 63073071-63073071 on Build GRCh38
rs772541661 Chr.8: 63072899-63072899 on Build GRCh38
rs375919660 Chr.8: 63073106-63073106 on Build GRCh38

View Full Product Details