Assay Design
Gene Transcripts
- Gene Symbol
- SMCR5
- Entrez Gene ID
-
140771
- Gene Name
- Smith-Magenis syndrome chromosome region, candidate 5 (non-protein coding)
- Gene Aliases
-
NCRNA00034
- Chromosome Location
-
Chr.17: 17776686 - 17779529 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024007.1 |
|
1 - 1 |
54 |
83 |
GenBank mRNA |
AF467442.1 |
- |
1 - 1 |
54 |
83 |
View Full Product Details