Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037325.2 | NP_001032402.1 | 2 - 3 | 473 | 84 |
NM_001199165.2 | NP_001186094.1 | 22 - 23 | 2604 | 84 | |
NM_001302891.1 | NP_001289820.1 | 20 - 21 | 2554 | 84 | |
XM_005257119.4 | XP_005257176.1 | 22 - 23 | 2698 | 84 | |
XM_005257125.3 | XP_005257182.1 | 14 - 15 | 1688 | 84 | |
XM_005257126.4 | XP_005257183.1 | 11 - 12 | 1353 | 84 | |
XM_006721740.2 | XP_006721803.1 | 22 - 23 | 2570 | 84 | |
XM_011524461.2 | XP_011522763.1 | 22 - 23 | 2711 | 84 | |
XM_011524462.2 | XP_011522764.1 | 22 - 23 | 2700 | 84 | |
XM_011524463.2 | XP_011522765.1 | 22 - 23 | 2700 | 84 | |
XM_011524465.2 | XP_011522767.1 | 19 - 20 | 2027 | 84 | |
XM_011524466.2 | XP_011522768.1 | 12 - 13 | 1506 | 84 | |
XM_011524467.2 | XP_011522769.1 | 11 - 12 | 1348 | 84 | |
XM_017024302.1 | XP_016879791.1 | 20 - 21 | 2441 | 84 | |
GenBank mRNA | AF458591.1 | - | 12 - 13 | 1511 | 84 |
AK299075.1 | - | 22 - 23 | 2555 | 84 | |
BC029524.1 | - | 2 - 3 | 473 | 84 | |
BC131805.1 | - | 20 - 21 | 2382 | 84 | |
BX097629.1 | - | 2 - 3 | 151 | 84 |