Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145038.4 | NP_659475.2 | 13 - 14 | 1768 | 60 |
XM_005264637.3 | XP_005264694.1 | 12 - 13 | 1576 | 60 | |
XM_005264638.3 | XP_005264695.1 | 9 - 10 | 1123 | 60 | |
XM_017005271.1 | XP_016860760.1 | 12 - 13 | 1536 | 60 | |
GenBank mRNA | AK057222.1 | - | 13 - 14 | 1764 | 60 |
AK289953.1 | - | 13 - 14 | 1751 | 60 | |
AL833892.1 | - | 12 - 13 | 1502 | 60 | |
BC009488.2 | - | 12 - 13 | 1491 | 60 |