Assay Design



Gene Transcripts

Gene Symbol
MAP4K4
Entrez Gene ID
9448
Gene Name
mitogen-activated protein kinase kinase kinase kinase 4
Gene Aliases
FLH21957, HEL-S-31, HGK, MEKKK4, NIK
Chromosome Location
Chr.2: 101697703 - 101894690 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242559.1 NP_001229488.1 9 - 10 781 65
NM_001242560.1 NP_001229489.1 9 - 10 1154 65
NM_004834.4 NP_004825.3 9 - 10 1154 65
NM_145686.3 NP_663719.2 9 - 10 1154 65
NM_145687.3 NP_663720.1 9 - 10 781 65
XM_005264048.2 XP_005264105.1 9 - 10 850 65
XM_005264050.3 XP_005264107.1 9 - 10 850 65
XM_005264052.3 XP_005264109.1 9 - 10 850 65
XM_005264057.3 XP_005264114.1 9 - 10 850 65
XM_005264058.3 XP_005264115.1 9 - 10 850 65
XM_005264059.3 XP_005264116.1 9 - 10 850 65
XM_005264060.3 XP_005264117.1 9 - 10 850 65
XM_005264062.3 XP_005264119.1 9 - 10 850 65
XM_005264063.3 XP_005264120.1 9 - 10 850 65
XM_005264064.3 XP_005264121.1 9 - 10 850 65
XM_005264065.3 XP_005264122.1 9 - 10 850 65
XM_005264066.3 XP_005264123.1 9 - 10 850 65
XM_005264068.3 XP_005264125.1 9 - 10 850 65
XM_005264069.2 XP_005264126.1 9 - 10 850 65
XM_005264071.2 XP_005264128.1 9 - 10 850 65
XM_006712865.2 XP_006712928.1 9 - 10 850 65
XM_006712868.2 XP_006712931.1 9 - 10 850 65
XM_006712869.2 XP_006712932.1 9 - 10 850 65
XM_017005349.1 XP_016860838.1 9 - 10 850 65
XM_017005350.1 XP_016860839.1 9 - 10 850 65
XM_017005351.1 XP_016860840.1 9 - 10 850 65
XM_017005352.1 XP_016860841.1 9 - 10 850 65
XM_017005353.1 XP_016860842.1 9 - 10 850 65
XM_017005354.1 XP_016860843.1 9 - 10 850 65
XM_017005355.1 XP_016860844.1 9 - 10 850 65
XM_017005356.1 XP_016860845.1 9 - 10 850 65
XM_017005357.1 XP_016860846.1 9 - 10 850 65
XM_017005358.1 XP_016860847.1 9 - 10 850 65
XM_017005359.1 XP_016860848.1 9 - 10 850 65
XM_017005360.1 XP_016860849.1 9 - 10 850 65
XM_017005361.1 XP_016860850.1 9 - 10 850 65
XM_017005362.1 XP_016860851.1 9 - 10 850 65
XM_017005363.1 XP_016860852.1 9 - 10 850 65
XM_017005364.1 XP_016860853.1 9 - 10 850 65
XM_017005365.1 XP_016860854.1 9 - 10 850 65
XM_017005366.1 XP_016860855.1 9 - 10 850 65
XM_017005367.1 XP_016860856.1 9 - 10 850 65
XM_017005368.1 XP_016860857.1 9 - 10 850 65
XM_017005369.1 XP_016860858.1 9 - 10 850 65
XM_017005370.1 XP_016860859.1 9 - 10 850 65
XM_017005371.1 XP_016860860.1 9 - 10 850 65
XM_017005372.1 XP_016860861.1 9 - 10 850 65
XM_017005373.1 XP_016860862.1 9 - 10 850 65
XM_017005374.1 XP_016860863.1 9 - 10 850 65
XM_017005375.1 XP_016860864.1 9 - 10 850 65
GenBank mRNA AB014587.1 - 7 - 8 665 65
AF096300.1 - 9 - 10 855 65
AK074900.1 - 5 - 6 387 65
AK295610.1 - 7 - 8 746 65
AK296396.1 - 9 - 10 850 65
AK307879.1 - 5 - 6 336 65
AY212247.1 - 9 - 10 776 65
EU794634.1 - 9 - 10 831 65

View Full Product Details