Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019106.5 | NP_061979.3 | 5 - 6 | 695 | 81 |
NM_145733.2 | NP_663786.2 | 5 - 6 | 695 | 81 | |
XM_017028862.1 | XP_016884351.1 | 6 - 7 | 2293 | 81 | |
GenBank mRNA | AB209152.1 | - | 2 - 3 | 162 | 81 |
AF285107.1 | - | 5 - 6 | 572 | 81 | |
AF285108.1 | - | 3 - 4 | 361 | 81 | |
AF285109.1 | - | 5 - 6 | 572 | 81 | |
AK290037.1 | - | 5 - 6 | 680 | 81 | |
AK299919.1 | - | 4 - 5 | 440 | 81 | |
AL833942.1 | - | 5 - 6 | 630 | 81 | |
BC111779.1 | - | 5 - 6 | 588 | 81 | |
BX465056.2 | - | 5 - 6 | 809 | 81 | |
CR456572.1 | - | 5 - 6 | 584 | 81 |