Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145754.3 | NP_665697.1 | 8 - 9 | 972 | 82 |
XM_005272357.3 | XP_005272414.1 | 8 - 9 | 1062 | 82 | |
XM_011517362.2 | XP_011515664.1 | 8 - 9 | 1063 | 82 | |
GenBank mRNA | AK055045.1 | - | 5 - 6 | 350 | 82 |
BC017311.2 | - | 9 - 10 | 954 | 82 |