Assay Design



Gene Transcripts

Gene Symbol
LGI2
Entrez Gene ID
55203
Gene Name
leucine rich repeat LGI family member 2
Gene Aliases
LGIL2
Chromosome Location
Chr.4: 24991960 - 25030792 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018176.3 NP_060646.2 3 - 4 439 95
XM_011513850.2 XP_011512152.1 3 - 4 341 95
XM_017008356.1 XP_016863845.1 3 - 4 341 95
GenBank mRNA AB067503.1 - 3 - 4 332 95
AF467955.1 - 3 - 4 340 95
AJ487516.1 - 3 - 4 340 95
AJ487958.1 - 3 - 4 362 95
AK126093.1 - 1 - 2 53 95
AL833647.1 - 2 - 3 130 95
BC101759.1 - 3 - 4 439 95
BC101761.1 - 3 - 4 439 95

View Full Product Details