Assay Design
Gene Transcripts
- Gene Symbol
- PANK1
- Entrez Gene ID
-
53354
- Gene Name
- pantothenate kinase 1
- Gene Aliases
-
PANK
- Chromosome Location
-
Chr.10: 89579497 - 89645572 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138316.3 |
NP_612189.2 |
3 - 4 |
803 |
86 |
GenBank mRNA |
AF355198.1 |
- |
3 - 4 |
787 |
86 |
View Full Product Details