Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077199.2 | NP_001070667.1 | 10 - 11 | 1740 | 117 |
NM_001323527.1 | NP_001310456.1 | 11 - 12 | 1864 | 117 | |
NM_001323534.1 | NP_001310463.1 | 11 - 12 | 1755 | 117 | |
NM_139168.3 | NP_631907.1 | 11 - 12 | 1893 | 117 | |
XM_011543171.2 | XP_011541473.1 | 10 - 11 | 1965 | 117 | |
XM_017009055.1 | XP_016864544.1 | 9 - 10 | 1274 | 117 | |
XM_017009056.1 | XP_016864545.1 | 10 - 11 | 1525 | 117 | |
GenBank mRNA | AF459094.1 | - | 10 - 11 | 1574 | 117 |
AK091758.1 | - | 10 - 11 | 1622 | 117 | |
AK125893.1 | - | 3 - 4 | 1425 | 117 | |
BC067770.1 | - | 11 - 12 | 1894 | 117 | |
BC112343.1 | - | 8 - 9 | 1262 | 117 |