Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083885.2 | NP_001077354.2 | 11 - 12 | 2031 | 60 |
NM_001173425.1 | NP_001166896.1 | 11 - 12 | 3192 | 60 | |
NM_015404.3 | NP_056219.3 | 11 - 12 | 3195 | 60 | |
XM_011518484.1 | XP_011516786.1 | 12 - 13 | 3281 | 60 | |
XM_011518485.1 | XP_011516787.1 | 13 - 14 | 3129 | 60 | |
XM_011518486.1 | XP_011516788.1 | 12 - 13 | 3278 | 60 | |
XM_011518487.1 | XP_011516789.1 | 11 - 12 | 3155 | 60 | |
XM_011518488.1 | XP_011516790.1 | 10 - 11 | 3038 | 60 | |
XM_011518495.1 | XP_011516797.1 | 9 - 10 | 1533 | 60 | |
GenBank mRNA | AB040959.1 | - | 11 - 12 | 2709 | 60 |
AK022854.1 | - | 7 - 8 | 1909 | 60 | |
AL110228.1 | - | 11 - 12 | 1989 | 60 | |
BC011918.2 | - | 2 - 3 | 304 | 60 | |
BC014524.1 | - | - | 3692 | 60 | |
BC136416.1 | - | 11 - 12 | 2941 | 60 | |
BC142614.1 | - | 11 - 12 | 3195 | 60 | |
BC142684.1 | - | 11 - 12 | 3170 | 60 |