Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013839.3 | NP_001013861.1 | 4 - 5 | 511 | 64 |
NM_001145297.3 | NP_001138769.1 | 4 - 5 | 511 | 64 | |
NM_001145298.3 | NP_001138770.1 | 4 - 5 | 511 | 64 | |
NM_001145299.3 | NP_001138771.1 | 4 - 5 | 511 | 64 | |
NM_001282313.1 | NP_001269242.1 | 3 - 4 | 501 | 64 | |
NM_015219.4 | NP_056034.2 | 4 - 5 | 511 | 64 | |
XM_006721786.3 | XP_006721849.1 | 4 - 5 | 510 | 64 | |
XM_006721787.3 | XP_006721850.1 | 4 - 5 | 945 | 64 | |
XM_006721788.3 | XP_006721851.1 | 4 - 5 | 510 | 64 | |
XM_006721789.3 | XP_006721852.1 | 4 - 5 | 510 | 64 | |
XM_011524571.2 | XP_011522873.1 | 2 - 3 | 290 | 64 | |
XM_011524572.2 | XP_011522874.1 | 4 - 5 | 510 | 64 | |
GenBank mRNA | AB028990.1 | - | 4 - 5 | 434 | 64 |
AK022397.1 | - | 4 - 5 | 467 | 64 | |
AK023832.1 | - | 4 - 5 | 419 | 64 | |
AK295871.1 | - | 3 - 4 | 453 | 64 | |
AY869728.1 | - | 2 - 3 | 280 | 64 | |
BC011045.1 | - | 4 - 5 | 445 | 64 | |
BC018466.1 | - | 4 - 5 | 419 | 64 | |
CB989148.1 | - | 4 - 5 | 526 | 64 | |
DA224278.1 | - | 4 - 5 | 472 | 64 | |
DC319923.1 | - | 3 - 4 | 501 | 64 | |
FJ457119.1 | - | 4 - 5 | 416 | 64 | |
FJ457120.1 | - | 4 - 5 | 416 | 64 |