Assay Design



Gene Transcripts

Gene Symbol
TPRG1L
Entrez Gene ID
127262
Gene Name
tumor protein p63 regulated 1-like
Gene Aliases
FAM79A, h-mover, mover
Chromosome Location
Chr.1: 3624992 - 3630131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182752.3 NP_877429.2 1 - 2 285 52
GenBank mRNA AK096427.1 - 1 - 2 262 52
AK289919.1 - 1 - 2 235 52
BC019034.2 - 1 - 2 227 52
BI547609.1 - 1 - 2 280 52

View Full Product Details