Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152721.5 | NP_689934.2 | 3 - 4 | 485 | 98 |
XM_011525875.2 | XP_011524177.1 | 3 - 4 | 506 | 98 | |
GenBank mRNA | AK057795.1 | - | 2 - 3 | 194 | 98 |
AY599248.1 | - | 3 - 4 | 483 | 98 | |
BC008583.1 | - | 1 - 2 | 67 | 98 | |
BC019045.2 | - | 1 - 2 | 110 | 98 | |
BC096744.1 | - | 3 - 4 | 478 | 98 |