Assay Design



Gene Transcripts

Gene Symbol
PYHIN1
Entrez Gene ID
149628
Gene Name
pyrin and HIN domain family member 1
Gene Aliases
IFIX
Chromosome Location
Chr.1: 158931177 - 159010083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005244930.1 XP_005244987.1 4 - 5 829 65
GenBank mRNA AK308350.1 - 4 - 5 809 65
BC020822.1 - 4 - 5 808 65

View Full Product Details