Assay Design
Gene Transcripts
- Gene Symbol
- CNTROB
- Entrez Gene ID
-
116840
- Gene Name
- centrobin, centriole duplication and spindle assembly protein
- Gene Aliases
-
LIP8, PP1221
- Chromosome Location
-
Chr.17: 7932054 - 7949919 on Build GRCh38
View Full Product Details