Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033200.2 | NP_149977.2 | 6 - 7 | 950 | 85 |
XM_005261955.3 | XP_005262012.1 | 6 - 7 | 1032 | 85 | |
XM_006724426.3 | XP_006724489.1 | 5 - 6 | 1396 | 85 | |
XM_006724427.3 | XP_006724490.1 | 6 - 7 | 947 | 85 | |
XM_017029077.1 | XP_016884566.1 | 6 - 7 | 1084 | 85 | |
GenBank mRNA | BC002942.2 | - | 6 - 7 | 924 | 85 |
BC014652.1 | - | 6 - 7 | 1013 | 85 | |
BC021143.1 | - | 6 - 7 | 924 | 85 | |
CR456605.1 | - | 6 - 7 | 934 | 85 |