Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127608.1 | NP_001121080.1 | 7 - 8 | 726 | 64 |
NM_004816.3 | NP_004807.3 | 7 - 8 | 1042 | 64 | |
XM_005252307.4 | XP_005252364.1 | 7 - 8 | 1603 | 64 | |
XM_017015324.1 | XP_016870813.1 | 6 - 7 | 787 | 64 | |
XM_017015325.1 | XP_016870814.1 | 7 - 8 | 726 | 64 | |
GenBank mRNA | AK056152.1 | - | 5 - 6 | 670 | 64 |
AK126127.1 | - | 7 - 8 | 1042 | 64 | |
BC021685.1 | - | 4 - 5 | 455 | 64 | |
BC113683.1 | - | 7 - 8 | 685 | 64 | |
BC113685.1 | - | 7 - 8 | 685 | 64 | |
BX641153.1 | - | 4 - 5 | 5895 | 64 |