Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166304.1 | NP_001159776.1 | 2 - 3 | 334 | 101 |
NM_017861.3 | NP_060331.3 | 2 - 3 | 334 | 101 | |
GenBank mRNA | AA879458.1 | - | 2 - 3 | 104 | 101 |
AK000529.1 | - | 2 - 3 | 297 | 101 | |
AK294340.1 | - | 2 - 3 | 517 | 101 | |
BC022542.1 | - | 2 - 3 | 294 | 101 |