Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178815.3 | NP_848930.1 | 4 - 5 | 590 | 123 |
XM_005252401.4 | XP_005252458.1 | 4 - 5 | 455 | 123 | |
GenBank mRNA | AF494061.1 | - | 4 - 5 | 512 | 123 |
AK027275.1 | - | 4 - 5 | 543 | 123 | |
BC024163.1 | - | 4 - 5 | 570 | 123 |