Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281710.1 | NP_001268639.1 | 9 - 10 | 1265 | 127 |
NM_001281711.1 | NP_001268640.1 | 9 - 10 | 1226 | 127 | |
NM_001281712.1 | NP_001268641.1 | 8 - 9 | 1045 | 127 | |
NM_015341.4 | NP_056156.2 | 9 - 10 | 1298 | 127 | |
XM_005263908.3 | XP_005263965.1 | 9 - 10 | 1238 | 127 | |
XM_006712388.3 | XP_006712451.1 | 9 - 10 | 1238 | 127 | |
GenBank mRNA | AK291053.1 | - | 9 - 10 | 1239 | 127 |
AK299248.1 | - | 9 - 10 | 1167 | 127 | |
AK303725.1 | - | 8 - 9 | 990 | 127 | |
AK308050.1 | - | 9 - 10 | 1205 | 127 | |
AK310649.1 | - | 9 - 10 | 1166 | 127 | |
BC024211.2 | - | 9 - 10 | 1252 | 127 | |
D38553.1 | - | 9 - 10 | 1228 | 127 |