Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173909474-173909986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINC1
Entrez Gene ID
462
Gene Name
serpin family C member 1
Gene Aliases
AT3, AT3D, ATIII, THPH7
UniGene
Hs.75599
Gene Chromosome Location
Chr.1: 173903804-173917378 on Build GRCh38
Transcripts
NM_000488.3 XM_005245198.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5735100 c.934A>G p.T312A Chr.1: 173909771-173909771 on Build GRCh38
899814 c.877C>T p.R293W Chr.1: 173909828-173909828 on Build GRCh38
2150092 c.1119C>T p.V373V Chr.1: 173909586-173909586 on Build GRCh38
677071 c.1015T>A p.W339R Chr.1: 173909690-173909690 on Build GRCh38
5788124 c.987G>A p.K329K Chr.1: 173909718-173909718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs483352856 Chr.1: 173909689-173909689 on Build GRCh38
rs377588972 Chr.1: 173909833-173909833 on Build GRCh38
rs747977353 Chr.1: 173909504-173909504 on Build GRCh38
rs192187532 Chr.1: 173909700-173909700 on Build GRCh38
rs779331710 Chr.1: 173909535-173909535 on Build GRCh38
rs755808418 Chr.1: 173909683-173909683 on Build GRCh38
rs549991084 Chr.1: 173909791-173909791 on Build GRCh38
rs1036017552 Chr.1: 173909838-173909838 on Build GRCh38
rs934275554 Chr.1: 173909538-173909538 on Build GRCh38
rs5877 Chr.1: 173909724-173909724 on Build GRCh38
rs886045594 Chr.1: 173909651-173909651 on Build GRCh38
rs778650224 Chr.1: 173909872-173909872 on Build GRCh38
rs909946904 Chr.1: 173909571-173909571 on Build GRCh38
rs959244848 Chr.1: 173909736-173909736 on Build GRCh38
rs774262708 Chr.1: 173909709-173909709 on Build GRCh38
rs572313182 Chr.1: 173909827-173909827 on Build GRCh38
rs745879490 Chr.1: 173909883-173909883 on Build GRCh38
rs746961120 Chr.1: 173909928-173909928 on Build GRCh38
rs758801372 Chr.1: 173909668-173909668 on Build GRCh38
rs770852197 Chr.1: 173909939-173909939 on Build GRCh38
rs150507232 Chr.1: 173909795-173909795 on Build GRCh38
rs762780270 Chr.1: 173909600-173909600 on Build GRCh38
rs776791729 Chr.1: 173909747-173909747 on Build GRCh38
rs766626880 Chr.1: 173909735-173909735 on Build GRCh38
rs745357314 Chr.1: 173909676-173909676 on Build GRCh38
rs780507091 Chr.1: 173909515-173909515 on Build GRCh38
rs780549243 Chr.1: 173909561-173909561 on Build GRCh38
rs1037223864 Chr.1: 173909532-173909532 on Build GRCh38
rs5878 Chr.1: 173909694-173909694 on Build GRCh38
rs758087836 Chr.1: 173909900-173909900 on Build GRCh38
rs184508490 Chr.1: 173909823-173909823 on Build GRCh38
rs761024265 Chr.1: 173909635-173909635 on Build GRCh38
rs762004419 Chr.1: 173909620-173909620 on Build GRCh38
rs773399107 Chr.1: 173909725-173909725 on Build GRCh38
rs759002598 Chr.1: 173909769-173909769 on Build GRCh38
rs756267601 Chr.1: 173909510-173909510 on Build GRCh38
rs200857896 Chr.1: 173909730-173909730 on Build GRCh38
rs745578147 Chr.1: 173909568-173909568 on Build GRCh38
rs781308217 Chr.1: 173909947-173909947 on Build GRCh38
rs765761813 Chr.1: 173909645-173909645 on Build GRCh38
rs567550044 Chr.1: 173909710-173909710 on Build GRCh38
rs906122970 Chr.1: 173909931-173909931 on Build GRCh38
rs373515340 Chr.1: 173909638-173909638 on Build GRCh38
rs960126155 Chr.1: 173909654-173909654 on Build GRCh38
rs747142328 Chr.1: 173909825-173909825 on Build GRCh38
rs777786232 Chr.1: 173909921-173909921 on Build GRCh38
rs868179720 Chr.1: 173909942-173909942 on Build GRCh38
rs483352857 Chr.1: 173909851-173909852 on Build GRCh38
rs984249696 Chr.1: 173909739-173909739 on Build GRCh38
rs121909565 Chr.1: 173909564-173909564 on Build GRCh38
rs764695432 Chr.1: 173909834-173909834 on Build GRCh38
rs145418597 Chr.1: 173909958-173909958 on Build GRCh38
rs759734009 Chr.1: 173909753-173909753 on Build GRCh38
rs979120581 Chr.1: 173909722-173909722 on Build GRCh38
rs775413132 Chr.1: 173909585-173909585 on Build GRCh38
rs1010218433 Chr.1: 173909667-173909667 on Build GRCh38
rs765632646 Chr.1: 173909764-173909764 on Build GRCh38
rs749956661 Chr.1: 173909778-173909778 on Build GRCh38
rs370190321 Chr.1: 173909835-173909835 on Build GRCh38
rs1035744320 Chr.1: 173909642-173909642 on Build GRCh38
rs777398332 Chr.1: 173909655-173909655 on Build GRCh38
rs776662899 Chr.1: 173909828-173909828 on Build GRCh38
rs587776397 Chr.1: 173909824-173909824 on Build GRCh38
rs750607605 Chr.1: 173909506-173909506 on Build GRCh38
rs769991153 Chr.1: 173909831-173909831 on Build GRCh38
rs565091601 Chr.1: 173909609-173909609 on Build GRCh38
rs372820797 Chr.1: 173909819-173909819 on Build GRCh38
rs770381981 Chr.1: 173909554-173909554 on Build GRCh38
rs199469509 Chr.1: 173909622-173909639 on Build GRCh38
rs775845871 Chr.1: 173909832-173909832 on Build GRCh38
rs758316459 Chr.1: 173909644-173909644 on Build GRCh38
rs201381904 Chr.1: 173909822-173909822 on Build GRCh38
rs774393305 Chr.1: 173909611-173909611 on Build GRCh38
rs483352855 Chr.1: 173909670-173909672 on Build GRCh38
rs780821947 Chr.1: 173909675-173909675 on Build GRCh38
rs749129128 Chr.1: 173909697-173909697 on Build GRCh38
rs753056696 Chr.1: 173909767-173909767 on Build GRCh38
rs767683540 Chr.1: 173909627-173909627 on Build GRCh38
rs369894034 Chr.1: 173909537-173909537 on Build GRCh38
rs972361914 Chr.1: 173909817-173909817 on Build GRCh38
rs121909561 Chr.1: 173909681-173909684 on Build GRCh38
rs753254708 Chr.1: 173909646-173909646 on Build GRCh38
rs376752370 Chr.1: 173909639-173909639 on Build GRCh38
rs750144360 Chr.1: 173909516-173909519 on Build GRCh38
rs753390084 Chr.1: 173909871-173909871 on Build GRCh38
rs759821949 Chr.1: 173909830-173909830 on Build GRCh38
rs121909560 Chr.1: 173909737-173909738 on Build GRCh38
rs531513129 Chr.1: 173909516-173909516 on Build GRCh38
rs758329697 Chr.1: 173909889-173909889 on Build GRCh38
rs778915564 Chr.1: 173909820-173909820 on Build GRCh38
rs139275128 Chr.1: 173909897-173909897 on Build GRCh38
rs149006854 Chr.1: 173909582-173909582 on Build GRCh38
rs764455436 Chr.1: 173909770-173909770 on Build GRCh38
rs551575090 Chr.1: 173909531-173909531 on Build GRCh38
rs139463995 Chr.1: 173909847-173909847 on Build GRCh38
rs773134244 Chr.1: 173909628-173909628 on Build GRCh38
rs863224495 Chr.1: 173909645-173909645 on Build GRCh38
rs745890007 Chr.1: 173909954-173909954 on Build GRCh38

View Full Product Details