Assay Design



Gene Transcripts

Gene Symbol
ZFP28
Entrez Gene ID
140612
Gene Name
ZFP28 zinc finger protein
Gene Aliases
mkr5
Chromosome Location
Chr.19: 56536791 - 56559213 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308440.1 NP_001295369.1 1 - 2 278 111
NM_020828.1 NP_065879.1 1 - 2 278 111
GenBank mRNA AB037852.1 - 1 - 2 278 111
AF507045.1 - 1 - 2 278 111
AK289583.1 - 1 - 2 263 111
BC028416.2 - 1 - 2 273 111
BC099903.1 - 1 - 2 273 111
BC127002.1 - 1 - 2 263 111
BC127003.1 - 1 - 2 263 111

View Full Product Details