Assay Design



Gene Transcripts

Gene Symbol
LHFPL5
Entrez Gene ID
222662
Gene Name
lipoma HMGIC fusion partner-like 5
Gene Aliases
DFNB67, TMHS, dJ510O8.8
Chromosome Location
Chr.6: 35805294 - 35824075 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182548.3 NP_872354.1 3 - 4 1056 136
GenBank mRNA AK126841.1 - 3 - 4 971 136
BC028630.2 - 3 - 4 1056 136

View Full Product Details