Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317721.1 | NP_001304650.1 | 3 - 4 | 653 | 94 |
NM_152892.2 | NP_690852.1 | 3 - 4 | 635 | 94 | |
XM_017011845.1 | XP_016867334.1 | 3 - 4 | 536 | 94 | |
GenBank mRNA | AB209379.1 | - | 3 - 4 | 513 | 94 |
AK290967.1 | - | 3 - 4 | 534 | 94 | |
AK313771.1 | - | 3 - 4 | 518 | 94 | |
AL133057.1 | - | 3 - 4 | 585 | 94 | |
BC009436.1 | - | 2 - 3 | 326 | 94 | |
BC018769.1 | - | 2 - 3 | 165 | 94 | |
BC030547.1 | - | 3 - 4 | 575 | 94 |