Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033402.4 | NP_208325.3 | 18 - 19 | 3183 | 87 |
XM_005251318.3 | XP_005251375.1 | 17 - 18 | 2972 | 87 | |
XM_005251319.4 | XP_005251376.1 | 16 - 17 | 2666 | 87 | |
XM_005251320.3 | XP_005251377.1 | 15 - 16 | 2458 | 87 | |
XM_005251322.3 | XP_005251379.1 | 14 - 15 | 2272 | 87 | |
XM_017013920.1 | XP_016869409.1 | 19 - 20 | 3131 | 87 | |
XM_017013921.1 | XP_016869410.1 | 19 - 20 | 3068 | 87 | |
XM_017013922.1 | XP_016869411.1 | 18 - 19 | 2945 | 87 | |
XM_017013923.1 | XP_016869412.1 | 18 - 19 | 2958 | 87 | |
XM_017013925.1 | XP_016869414.1 | 16 - 17 | 2677 | 87 | |
XM_017013926.1 | XP_016869415.1 | 16 - 17 | 2660 | 87 | |
XM_017013927.1 | XP_016869416.1 | 17 - 18 | 2688 | 87 | |
XM_017013928.1 | XP_016869417.1 | 14 - 15 | 2140 | 87 | |
GenBank mRNA | AB051551.1 | - | 17 - 18 | 3804 | 87 |
AK057990.1 | - | 9 - 10 | 1438 | 87 | |
AK302648.1 | - | 18 - 19 | 3033 | 87 | |
BC030701.2 | - | 18 - 19 | 3056 | 87 |