Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152431.2 | NP_689644.2 | 17 - 18 | 2384 | 64 |
GenBank mRNA | AB079366.1 | - | 17 - 18 | 2344 | 64 |
AK096837.1 | - | 13 - 14 | 1642 | 64 | |
BC031060.1 | - | 17 - 18 | 2176 | 64 | |
CR749642.1 | - | 16 - 17 | 2638 | 64 |