Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005272.3 | NP_005263.1 | 4 - 5 | 668 | 83 |
XM_011541264.2 | XP_011539566.1 | 5 - 6 | 759 | 83 | |
GenBank mRNA | AF493909.1 | - | 4 - 5 | 455 | 83 |
BC000233.1 | - | 4 - 5 | 636 | 83 | |
BT009844.1 | - | 4 - 5 | 455 | 83 |