Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005272.3 | NP_005263.1 | 6 - 7 | 935 | 99 |
XM_011541264.2 | XP_011539566.1 | 7 - 8 | 1026 | 99 | |
GenBank mRNA | AF493909.1 | - | 6 - 7 | 722 | 99 |
BC000233.1 | - | 6 - 7 | 903 | 99 | |
BT009844.1 | - | 6 - 7 | 722 | 99 |