Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021136.2 | NP_066959.1 | 8 - 9 | 2496 | 71 |
NM_206852.2 | NP_996734.1 | 6 - 7 | 891 | 71 | |
XM_017021578.1 | XP_016877067.1 | 6 - 7 | 780 | 71 | |
GenBank mRNA | AK128113.1 | - | 3 - 4 | 2356 | 71 |
AK290534.1 | - | 6 - 7 | 891 | 71 | |
AK297492.1 | - | 7 - 8 | 798 | 71 | |
BC000314.2 | - | 6 - 7 | 676 | 71 | |
BC003003.1 | - | 6 - 7 | 676 | 71 | |
BC090862.1 | - | 8 - 9 | 2411 | 71 | |
BC111694.1 | - | 9 - 10 | 2186 | 71 | |
BX449967.2 | - | 6 - 7 | 879 | 71 | |
CR457030.1 | - | 6 - 7 | 583 | 71 | |
L10333.1 | - | 8 - 9 | 2409 | 71 | |
L10334.1 | - | 6 - 7 | 1317 | 71 | |
L10335.1 | - | 6 - 7 | 611 | 71 |