Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024100.3 | NP_077005.2 | 2 - 3 | 349 | 65 |
XM_017027027.1 | XP_016882516.1 | 4 - 5 | 875 | 65 | |
GenBank mRNA | BC000040.2 | - | 2 - 3 | 334 | 65 |
BC001648.2 | - | 2 - 3 | 329 | 65 | |
CB991338.1 | - | - | 376 | 65 |