Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203462.2 | NP_982287.1 | 1 - 2 | 646 | 89 |
GenBank mRNA | AF155654.1 | - | 1 - 2 | 642 | 89 |
BC008087.1 | - | 1 - 2 | 593 | 89 | |
BC066897.1 | - | 1 - 2 | 646 | 89 | |
BX397546.2 | - | 1 - 2 | 89 |