Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77199773-77200172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS13A
Entrez Gene ID
23230
Gene Name
vacuolar protein sorting 13 homolog A
Gene Aliases
CHAC, CHOREIN
UniGene
Hs.459790
Gene Chromosome Location
Chr.9: 77177353-77421537 on Build GRCh38
Transcripts
NM_001018037.1 NM_001018038.2 NM_015186.3 NM_033305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5411314 c.110C>T p.A37V Chr.9: 77199954-77199954 on Build GRCh38
5935529 c.144+1G>A p.? Chr.9: 77199989-77199989 on Build GRCh38
4740404 c.117G>T p.K39N Chr.9: 77199961-77199961 on Build GRCh38
6820154 c.143T>C p.L48P Chr.9: 77199987-77199987 on Build GRCh38
5411315 c.110C>T p.A37V Chr.9: 77199954-77199954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769097566 Chr.9: 77199936-77199936 on Build GRCh38
rs762167465 Chr.9: 77199941-77199941 on Build GRCh38
rs960135788 Chr.9: 77199960-77199960 on Build GRCh38
rs746374841 Chr.9: 77200088-77200088 on Build GRCh38
rs527950248 Chr.9: 77199870-77199870 on Build GRCh38
rs747022718 Chr.9: 77199799-77199801 on Build GRCh38
rs140518771 Chr.9: 77199970-77199970 on Build GRCh38
rs570109947 Chr.9: 77200026-77200026 on Build GRCh38
rs1051397652 Chr.9: 77199922-77199922 on Build GRCh38
rs912902372 Chr.9: 77200110-77200110 on Build GRCh38
rs895396539 Chr.9: 77200147-77200147 on Build GRCh38
rs921749304 Chr.9: 77200068-77200068 on Build GRCh38
rs773656342 Chr.9: 77199968-77199968 on Build GRCh38
rs1031301708 Chr.9: 77199888-77199888 on Build GRCh38
rs971120184 Chr.9: 77199881-77199881 on Build GRCh38
rs778907324 Chr.9: 77200008-77200008 on Build GRCh38
rs957530886 Chr.9: 77199916-77199916 on Build GRCh38
rs1025656538 Chr.9: 77199985-77199985 on Build GRCh38
rs1043664164 Chr.9: 77199932-77199932 on Build GRCh38
rs757929970 Chr.9: 77200021-77200021 on Build GRCh38
rs749639590 Chr.9: 77199845-77199848 on Build GRCh38
rs963255421 Chr.9: 77200025-77200025 on Build GRCh38
rs12552898 Chr.9: 77199883-77199883 on Build GRCh38
rs746589360 Chr.9: 77200034-77200034 on Build GRCh38
rs763917022 Chr.9: 77200015-77200016 on Build GRCh38
rs1048204621 Chr.9: 77200100-77200100 on Build GRCh38
rs942010006 Chr.9: 77200109-77200109 on Build GRCh38
rs990061243 Chr.9: 77199928-77199928 on Build GRCh38
rs202024224 Chr.9: 77199931-77199931 on Build GRCh38
rs983390862 Chr.9: 77200010-77200010 on Build GRCh38
rs372018749 Chr.9: 77199973-77199973 on Build GRCh38
rs758443738 Chr.9: 77199936-77199937 on Build GRCh38
rs768368639 Chr.9: 77200038-77200038 on Build GRCh38
rs752636187 Chr.9: 77199937-77199937 on Build GRCh38
rs933085486 Chr.9: 77200084-77200087 on Build GRCh38
rs368914910 Chr.9: 77199962-77199962 on Build GRCh38
rs753683797 Chr.9: 77200002-77200002 on Build GRCh38
rs376952243 Chr.9: 77199982-77199982 on Build GRCh38
rs933055967 Chr.9: 77199908-77199908 on Build GRCh38
rs764893746 Chr.9: 77199925-77199925 on Build GRCh38
rs750432248 Chr.9: 77200017-77200017 on Build GRCh38
rs185381432 Chr.9: 77199839-77199839 on Build GRCh38
rs538776323 Chr.9: 77200097-77200097 on Build GRCh38
rs909190503 Chr.9: 77200101-77200101 on Build GRCh38
rs1055333881 Chr.9: 77200143-77200143 on Build GRCh38
rs535834952 Chr.9: 77199882-77199882 on Build GRCh38
rs182861770 Chr.9: 77200003-77200003 on Build GRCh38
rs770319746 Chr.9: 77199947-77199947 on Build GRCh38
rs35470636 Chr.9: 77200057-77200058 on Build GRCh38
rs779738921 Chr.9: 77200022-77200022 on Build GRCh38
rs974574402 Chr.9: 77200064-77200064 on Build GRCh38
rs375087579 Chr.9: 77200035-77200035 on Build GRCh38
rs763733189 Chr.9: 77199984-77199984 on Build GRCh38
rs190684534 Chr.9: 77199938-77199938 on Build GRCh38

View Full Product Details