Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77212837-77213366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS13A
Entrez Gene ID
23230
Gene Name
vacuolar protein sorting 13 homolog A
Gene Aliases
CHAC, CHOREIN
UniGene
Hs.459790
Gene Chromosome Location
Chr.9: 77177353-77421537 on Build GRCh38
Transcripts
NM_001018037.1 NM_001018038.2 NM_015186.3 NM_033305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1110212 c.557C>T p.T186I Chr.9: 77212970-77212970 on Build GRCh38
5962119 c.688A>T p.N230Y Chr.9: 77213306-77213306 on Build GRCh38
4636189 c.690C>T p.N230N Chr.9: 77213308-77213308 on Build GRCh38
1110213 c.557C>T p.T186I Chr.9: 77212970-77212970 on Build GRCh38
456099 c.600G>C p.E200D Chr.9: 77213013-77213013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779916647 Chr.9: 77213308-77213308 on Build GRCh38
rs1055696669 Chr.9: 77213115-77213115 on Build GRCh38
rs192380515 Chr.9: 77213001-77213001 on Build GRCh38
rs376856665 Chr.9: 77213053-77213053 on Build GRCh38
rs767393485 Chr.9: 77212951-77212952 on Build GRCh38
rs770388865 Chr.9: 77213050-77213050 on Build GRCh38
rs908278736 Chr.9: 77212937-77212937 on Build GRCh38
rs754716705 Chr.9: 77213319-77213319 on Build GRCh38
rs865951534 Chr.9: 77213268-77213268 on Build GRCh38
rs375793684 Chr.9: 77213068-77213068 on Build GRCh38
rs777737006 Chr.9: 77213242-77213242 on Build GRCh38
rs752509925 Chr.9: 77213023-77213023 on Build GRCh38
rs772782295 Chr.9: 77212923-77212923 on Build GRCh38
rs926044207 Chr.9: 77213093-77213093 on Build GRCh38
rs765321686 Chr.9: 77213216-77213216 on Build GRCh38
rs750602609 Chr.9: 77213226-77213226 on Build GRCh38
rs573841519 Chr.9: 77213206-77213206 on Build GRCh38
rs748237913 Chr.9: 77213186-77213187 on Build GRCh38
rs763166254 Chr.9: 77213231-77213231 on Build GRCh38
rs17723908 Chr.9: 77213048-77213048 on Build GRCh38
rs979287409 Chr.9: 77213079-77213079 on Build GRCh38
rs761927584 Chr.9: 77213211-77213211 on Build GRCh38
rs571798246 Chr.9: 77213101-77213101 on Build GRCh38
rs964649572 Chr.9: 77213279-77213279 on Build GRCh38
rs372228857 Chr.9: 77213007-77213007 on Build GRCh38
rs146941948 Chr.9: 77213239-77213239 on Build GRCh38
rs199999368 Chr.9: 77212940-77212940 on Build GRCh38
rs1030540170 Chr.9: 77213324-77213324 on Build GRCh38
rs761860828 Chr.9: 77212983-77212983 on Build GRCh38
rs755223567 Chr.9: 77212959-77212960 on Build GRCh38
rs765411702 Chr.9: 77212994-77212994 on Build GRCh38
rs796155513 Chr.9: 77212978-77212978 on Build GRCh38
rs771869402 Chr.9: 77213281-77213281 on Build GRCh38
rs1005003361 Chr.9: 77212878-77212878 on Build GRCh38
rs567123315 Chr.9: 77212889-77212889 on Build GRCh38
rs376299838 Chr.9: 77212928-77212928 on Build GRCh38
rs886198919 Chr.9: 77212877-77212877 on Build GRCh38
rs760999983 Chr.9: 77213186-77213186 on Build GRCh38
rs778895810 Chr.9: 77213030-77213033 on Build GRCh38
rs1017574389 Chr.9: 77213254-77213254 on Build GRCh38
rs556470398 Chr.9: 77213257-77213257 on Build GRCh38
rs769146114 Chr.9: 77212919-77212919 on Build GRCh38
rs745456320 Chr.9: 77213274-77213274 on Build GRCh38
rs759949545 Chr.9: 77212939-77212939 on Build GRCh38
rs529573724 Chr.9: 77212916-77212916 on Build GRCh38
rs761279676 Chr.9: 77212960-77212960 on Build GRCh38
rs1045643238 Chr.9: 77213094-77213094 on Build GRCh38
rs898898482 Chr.9: 77213336-77213336 on Build GRCh38
rs753770438 Chr.9: 77213245-77213245 on Build GRCh38
rs750690177 Chr.9: 77213000-77213000 on Build GRCh38
rs777630056 Chr.9: 77213039-77213039 on Build GRCh38
rs17195354 Chr.9: 77213151-77213151 on Build GRCh38
rs1016014574 Chr.9: 77212883-77212883 on Build GRCh38
rs547102692 Chr.9: 77212888-77212888 on Build GRCh38
rs776004803 Chr.9: 77213080-77213080 on Build GRCh38
rs539259686 Chr.9: 77213092-77213092 on Build GRCh38
rs10869912 Chr.9: 77212875-77212875 on Build GRCh38
rs929454238 Chr.9: 77212899-77212899 on Build GRCh38
rs774215297 Chr.9: 77212935-77212935 on Build GRCh38
rs758528794 Chr.9: 77213331-77213334 on Build GRCh38
rs367896041 Chr.9: 77213059-77213059 on Build GRCh38
rs753936313 Chr.9: 77212962-77212962 on Build GRCh38
rs374507135 Chr.9: 77213209-77213209 on Build GRCh38
rs776094371 Chr.9: 77212948-77212948 on Build GRCh38
rs967569110 Chr.9: 77213075-77213075 on Build GRCh38
rs780994096 Chr.9: 77213016-77213016 on Build GRCh38
rs987663575 Chr.9: 77213063-77213063 on Build GRCh38
rs998694937 Chr.9: 77213119-77213119 on Build GRCh38
rs745540043 Chr.9: 77213054-77213054 on Build GRCh38
rs201208791 Chr.9: 77213241-77213241 on Build GRCh38
rs778600844 Chr.9: 77213270-77213270 on Build GRCh38
rs997728613 Chr.9: 77213304-77213304 on Build GRCh38
rs756091546 Chr.9: 77213024-77213024 on Build GRCh38
rs747364921 Chr.9: 77213318-77213318 on Build GRCh38
rs534170631 Chr.9: 77213126-77213126 on Build GRCh38
rs119477053 Chr.9: 77213240-77213240 on Build GRCh38

View Full Product Details