Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015329.3 | NP_056144.3 | 17 - 18 | 1675 | 70 |
XM_005259837.3 | XP_005259894.2 | 17 - 18 | 1664 | 70 | |
XM_005259838.3 | XP_005259895.3 | 15 - 16 | 1726 | 70 | |
XM_011527844.2 | XP_011526146.1 | 17 - 18 | 2080 | 70 | |
XM_017026539.1 | XP_016882028.1 | 15 - 16 | 1722 | 70 | |
GenBank mRNA | AB020699.1 | - | 17 - 18 | 1661 | 70 |
AK001349.1 | - | 5 - 6 | 392 | 70 | |
AK025896.1 | - | 15 - 16 | 1327 | 70 | |
AK125914.1 | - | 2 - 3 | 1894 | 70 | |
AK126227.1 | - | 4 - 5 | 1567 | 70 | |
AK128029.1 | - | 8 - 9 | 1659 | 70 | |
AL110250.1 | - | 6 - 7 | 450 | 70 | |
BC010222.1 | - | 6 - 7 | 461 | 70 | |
BC063863.1 | - | 6 - 7 | 808 | 70 |