Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006616.2 | NP_001006617.2 | 1 - 2 | 312 | 78 |
NM_012171.2 | NP_036303.1 | 1 - 2 | 312 | 78 | |
NM_130465.4 | NP_569732.2 | 1 - 2 | 312 | 78 | |
GenBank mRNA | AF174603.1 | - | 1 - 2 | 119 | 78 |
AK024427.1 | - | 1 - 2 | 202 | 78 | |
BC010405.2 | - | 1 - 2 | 134 | 78 | |
BC067105.1 | - | 1 - 2 | 197 | 78 | |
BI756001.1 | - | 1 - 2 | 274 | 78 |